Learn More
Medchemexpress LLC Simotinib hydrochloride | 944258-89-3 | 99.4% | 500.95 | C25H26ClFN4O4 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY101820100MG
Encompass_Preferred
Simotinib is a selective, orally bioavailable epidermal growth factor receptor (EGFR) tyrosine kinase inhibitor provided for research and preclinical studies. It exhibits antineoplastic activity by inhibiting EGFR (IC50 19.9 nM) and is supplied as a high-purity analytical standard for laboratory use.
  • Selective EGFR tyrosine kinase inhibitor.
  • Potent inhibition of EGFR (IC50 19.9 nM).
  • Orally bioavailable small molecule suitable for in vitro and preclinical work.
  • High purity (99.4%).
  • Available in multiple package sizes, including 100 mg.
  • Useful for oncology and EGFR signaling research applications.
Catalog No. 50-002-48566