Learn More
Novus Biologicals Simple Plex Control for Human CXCL12/SDF-1 alpha
SDP
Supplier:  Novus Biologicals 898082
Encompass
INTENDED USE AND DESCRIPTION: For use a quantitative control for the determination of human CXCL12/SDF-1 alpha concentrations in biological fluids. Controls are supplied lyophilized. Please refer to the lot specific certificate of analysis and Simple Plex Lyophilized Quality Control product insert for additional information.CXCL12/Stromal Cell-derived Factor 1 (SDF-1) is a member of the CXC chemokine ligand superfamily. Six alternatively spliced CXCL12/SDF-1 isoforms have been identified including, CXCL12/SDF-1 alpha, CXCL12/SDF-1 beta, and CXCL12/SDF-1 gamma. CXCL12/SDF-1 alpha is 89 amino acids (aa) in length with a predicted molecular weight of 10 kDa, CXCL12/SDF-1 beta is 93 aa in length with a predicted molecular weight of 10.6 kDa, and CXCL12/SDF-1 gamma is 119 aa in length with a predicted molecular weight of 13.7 kDa. The mouse CXCL12/SDF-1 orthologs share 99% aa sequence identity with the human protein.
Catalog No. NC3584897