Learn More
Novus Biologicals Simple Plex Control for Human VEGF
SDP
Supplier:  Novus Biologicals 894978
Encompass
INTENDED USE AND DESCRIPTION: For use a quantitative control for the determination of human VEGF concentrations in biological fluids. Controls are supplied lyophilized. Please refer to the lot specific certificate of analysis and Simple Plex Lyophilized Quality Control product insert for additional information.VEGF (Vascular endothelial growth factor) is a potent mediator of both angiogenesis and vasculogenesis in the fetus and adult. It is required for regulating the proliferation, migration, and survival of embryonic endothelial cells and during wound healing and the female reproductive cycle in adults. Pathologically, it is involved in tumor development and tumor vascular leakage. VEGF binds to VEGF R1/Flt-1, VEGF R2/Flk-1/KDR, and Neuropilin-1. Multiple forms of VEGF are generated by alternative splicing and proteolysis.
Catalog No. NC3584898