Learn More
Novus Biologicals Simple Plex Human CCL4/MIP-1 beta Cartridge.
SDP
Supplier:  Novus Biologicals SPCKBPS000359
Encompass
Simple Plex assay for the detection of human MIP-1 beta in cell culture supernatant (CCS), serum and plasma (EDTA/Heparin).
Catalog No. NC3235865