Learn More
Novus Biologicals Simple Plex Human IFN-gamma (3rd Gen) Cartridge
SDP
Supplier:  Novus Biologicals SPCKBPS002574
Encompass
Simple Plex assay for the detection of human IFN-gamma in cell culture supernatant (CCS), serum, plasma (EDTA/Heparin), and cerebrospinal fluid (CSF) on Ella automated benchtop ELISA.
Catalog No. NC3235874