Learn More
Novus Biologicals Simple Plex Human IL-6 3rd Gen Cartridge 1 Kit (1 Analyte x 72 Samples) /
SDP
Supplier:  Novus Biologicals SPCKBPS009384
Encompass
Simple Plex assay for the detection of human Interleukin 6 (IL-6) in cell culture supernatant (CCS), serum, plasma (EDTA/Heparin), saliva, urine, bronchoalveolar fluid (BALF), and cerebrospinal fluid (CSF).
Catalog No. NC3235876