Learn More
Novus Biologicals Simple Plex Human Serpin A1 Cartridge
SDP
Supplier:  Novus Biologicals SPCKBPS001047
Encompass
Simple Plex assay for the detection of human Serpin A1/ alpha 1-Antitrypsin in cell culture supernates (CCS), serum, plasma (EDTA/Heparin), urine, and cerebrospinal fluid (CSF).
Catalog No. NC3235870