Learn More
ProteinSimple Simple Plex Human SP-D Cartridge
Supplier:  ProteinSimple SPCKBPS000529
Encompass
Simple Plex assay for the detection of human Surfactant Protein D (SP-D) cell culture supernatant (CCS), serum and plasma (EDTA/Heparin).
Catalog No. NC3235869