Learn More
Novus Biologicals Simple Plex Human TREM-2 Cartridge
SDP
Supplier:  Novus Biologicals SPCKBPS001847
Encompass
Simple Plex assay for the detection of human TREM-2 in cell culture supernatant (CCS), serum, plasma (EDTA/Heparin), and cerebrospinal fluid (CSF).
Catalog No. NC3235873