Learn More
ProteinSimple SIMPLE PLX PRO A KIT 16 SAMPLE
Supplier:  ProteinSimple SPCKAOT009204
Encompass
Simple Plex Protein A assay kit contains cartridge, sample diluent SD62, and wash buffer - 1 Kit (1 Analyte x 16 Samples)
Catalog No. NC3521450