Learn More
BANGS LABORATORIES INC Simply Cellular Anti-Human IgG Microspheres, Quantitative Analysis, Flow Cytometry, 5 mL
Supplier:  BANGS LABORATORIES INC 812B5ML
Encompass
Simply Cellular anti-Human IgG microspheres provide a reliable standard for quantitative analysis in flow cytometry, allowing researchers to effectively determine cellular antigen expression and establish Antibody Binding Capacity (ABC) values. Designed with a single population of microspheres coated with a specific capture antibody for human IgG, their size closely resembles that of human lymphocytes, enhancing the relevance of cellular assays. With a concentration of 2e+6 beads/mL, they enable extensive testing capabilities.
Catalog No. NC0306499