Learn More
BANGS LABORATORIES INC Simply Cellular Anti-Mouse IgG, Flow Cytometry Standard, Quality Control Reagent, 5 mL
Supplier:  BANGS LABORATORIES INC 810B5ML
Encompass
Simply Cellular anti-Mouse IgG is a specialized flow cytometry standard designed for labeling monoclonal IgG antibodies, providing researchers with essential tools to assess the fluorescence intensity and stability of conjugated antibodies. Its microspheres offer precise fluorophore-to-protein (F/P) ratio determination and serve as effective quality control reagents. With a volume of 5 mL, it's tailored for research use only, ensuring reliability in experimental applications.
Catalog No. NC0383512