Learn More
BANGS LABORATORIES INC Simply Cellular Anti-Mouse IgG, Flow Cytometry Standard, Fc-Specific Capture IgG, 1 mL
Supplier:  BANGS LABORATORIES INC 810A1ML
Encompass
The Simply Cellular anti-Mouse IgG is a flow cytometry standard designed for precise quantitative analysis of cellular antigen expression. Its uniform, cell-sized microspheres feature Fc-specific capture IgG, enabling effective assessment of antibody binding capacity and fluorophore-to-protein ratios. This product ensures consistent fluorescence measurements across experiments and is compatible with various staining methods.
Catalog No. NC9089181