Learn More
TARGETMOL CHEMICALS INC Simufilam dihydrochloride 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T91725MG
Encompass_Preferred
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. Simufilam dihydrochloride (PTI-125 dihydrochloride) is a low toxicity, orally active filamin A (FLNA) activator, which can be used for the research of Alzheimer's disease. Simufilam dihydrochloride preferentially binds altered FLNA and restores its native conformation, restoring receptor and synaptic activities and reducing its alpha7nAChR/TLR4 associations and downstream pathologies.
Catalog No. 50-287-7960