Learn More
Medchemexpress LLC Simufilam hydrochloride | 2480226-07-9 | 99.9% | 295.81 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY139142B50MG
Encompass_Preferred
Simufilam hydrochloride (PTI-125 hydrochloride) is an orally active FLNA modulator. It restores NMDAR signaling and Arc expression, inhibits overactive mTOR signaling, improves insulin sensitivity, and reduces Aβ42-induced neuroinflammation and tau protein hyperphosphorylation, making it suitable for Alzheimer's disease research.
  • Restores NMDAR signaling and Arc expression
  • Inhibits overactive mTOR signaling
  • Improves insulin sensitivity
  • Reduces Aβ42-induced neuroinflammation
  • Reduces tau protein hyperphosphorylation
  • Reduces FLNA-α7nAChR/TLR4 associations
  • Restores α7nAChR/NMDAR/IR function
  • Enhances spatial/working memory
Catalog No. 50-003-50806