Learn More
Enzo Life Sciences Simvastatin (50mg). CAS: 79902-63-9
SDP
Supplier:  Enzo Life Sciences BMLG2440050
Encompass_Preferred
HMG-CoA reductase inhibitor. Synthetic analog of Lovastatin (BML-G226). Inhibits the biosynthesis of isoprenoids and prevents small G protein activation. Displays lipid lowering and antihypertensive effects. Displays antiinflammatory, immunomodulatory effects as well as protective effects in experimental traumatic brain injury. Activates Rac1-dependent signalling pathway. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (>25mg/ml) or 100% ethanol (>25mg/ml); insoluble in water. Long Term Storage: -20°C.
Catalog No. 50-201-0875