Learn More
Medchemexpress LLC Simvastatin acid calcium hydrate | 530112-57-3 | >95.0% | 1 MG
SDP
Supplier:  Medchemexpress LLC HY119695B1MG
Encompass_Preferred
Simvastatin acid calcium hydrate (Tenivastatin) is the calcium hydrate salt of simvastatin acid, supplied as a research reagent and used in biochemical and cellular studies as an HMG-CoA reductase inhibitor.
  • Potent HMG-CoA reductase inhibitor for biochemical and cellular assays.
  • Reduces indoxyl sulfate-mediated reactive oxygen species in cardiomyocytes.
  • Modulates OATP3A1 expression in cardiomyocytes and HEK293 cells.
  • Offered in small research quantities suitable for assay development.
  • For research use only; not for human or clinical use.
Catalog No. 50-002-33001