Learn More
Medchemexpress LLC Simvastatin-d6 | 1002347-71-8 | 99.6% | 424.60 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY110231500UG
Encompass_Preferred
Simvastatin-d6 is a deuterium-labeled form of Simvastatin, which acts as a competitive inhibitor of HMG-CoA reductase with a Kᵢ of 0.2 nM. The incorporation of stable heavy isotopes into drug molecules makes it useful as a tracer for quantitative analysis during drug development. Deuteration has the potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer.
  • Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Incorporates stable heavy isotopes for quantitation during drug development.
  • Deuteration may impact pharmacokinetic and metabolic profiles of drugs.
  • Functions as a competitive inhibitor of HMG-CoA reductase.
  • Exhibits a high affinity with a Kᵢ of 0.2 nM for HMG-CoA reductase.
Catalog No. 50-003-44753