Learn More
Andwin Scientific Simzo Forehead Thermometers, Infrared Digital Thermometer for Baby Kids and Adults, Accurate Digital Reading Non-Contact and LCD Colorful Display
SDP
Supplier:  Andwin Scientific 990000584
Encompass_Preferred
The non-contact forehead thermometer is equipped with the latest medical infrared technology with 0-8cm effective measurement distance(5-8cm is more effective). You just need to pressing a button when you using this forehead digital thermometer, then it will provides accurate temperature instantly within 1 second. The easy-to-read LED display provides temperature readings in 3 colored backlist with green, orange or red to quickly identify a fever in the dark. If the temperature is on the normal range(95.9℉-99.1℉/35.5-37.3℃),the LCD will show green. If the temperature is above the normal range (default setting of 99.1℉ / 37.3℃), the fever thermometer will emit a warning sound and the LCD will show yellow or red.
Catalog No. NC1809250
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.