Learn More
Medchemexpress LLC Sinapaldehyde | 4206-58-0 | 208.21 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYN131250MG
Encompass_Preferred
Sinapaldehyde is a methoxyphenol extracted from various plants, functioning as a selective COX-2 inhibitor. It demonstrates antibacterial, anti-inflammatory, and antioxidant properties, making it suitable for various research applications.
  • Selective COX-2 inhibitor (IC50: 47.8 μM)
  • Scavenges DPPH free radicals (IC50: 172 μM)
  • Effective against Gram-negative and Gram-positive bacteria, including MRSA
  • Inhibits generation of NO and ROS
  • Reduces production of IL-6 and TNF-α
  • Inhibits expression of iNOS and COX-2
  • Inhibits phosphorylation of p65, ERK1/2, and SAPK/JNK
  • Solid appearance, white to yellow color
  • Soluble in DMSO (50 mg/mL)
Catalog No. 50-004-16283