Learn More
Apexbio Technology LLC Sinapinic acid 530-59-6 100mg
SDP
Supplier:  Apexbio Technology LLC C6649100
Encompass_Preferred
Sinapinic acid (CAS 530-59-6) is an organic compound commonly utilized as a matrix in matrix-assisted laser desorption/ionization (MALDI) mass spectrometry where it facilitates the desorption and ionization of proteins and peptides for accurate molecular weight determination The compound s ability to absorb laser energy and promote analyte ionization underpins its effectiveness in proteomics research Additionally sinapinic acid is naturally occurring in propolis contributing to the compound s significance in natural product studies Its primary research applications center on biomolecular analysis and the characterization of protein mixtures
Catalog No. 50-000-89218