Learn More
Medchemexpress LLC Sinapinic acid | 530-59-6 | 99.9% | 224.21 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYW00973210MM/1ML
Encompass_Preferred
Sinapinic acid is a phenolic compound isolated from Hydnophytum formicarum Jack. Rhizome. It acts as an inhibitor of HDAC with an IC50 of 2.27 mM, and also inhibits ACE-I activity. Sinapinic acid possesses potent anti-tumor activity, inducing apoptosis of tumor cells. It also demonstrates antioxidant and antidiabetic activities, reducing total cholesterol, triglyceride, and HOMA-IR index, and normalizing some serum parameters of antioxidative abilities and oxidative damage in ovariectomized rats.
  • Inhibitor of HDAC (IC50 of 2.27 mM).
  • Inhibits ACE-I activity.
  • Possesses potent anti-tumor activity and induces apoptosis of tumor cells.
  • Shows antioxidant activities.
  • Exhibits antidiabetic activities, including reducing total cholesterol, triglyceride, and HOMA-IR index.
  • Normalizes serum parameters related to antioxidative abilities and oxidative damage in ovariectomized rats.
Catalog No. 50-004-53329