Learn More
ARBOR SCIENTIFIC SINGING ROD
SDP
Supplier:  ARBOR SCIENTIFIC P77250
Encompass
Singing Rods make a great addition to your lessons on sound and waves. Like the Singing Sword of legend, these magical rods will sing for you...if you know the secret. Simply hold a rod between your thumb and forefinger at one of the three marked nodes.
Catalog No. NC0368888