Learn More
LICOR TBS Blocking Buffer and PVDF Kits (single buffer kit)
Supplier:  LICOR 92632099
Encompass_Preferred
The non-mammalian Intercept Blocker in these kits is in TBS (Tris-buffered saline) containing 0.06% Kathon II. The Immobilon-FL PVDF membrane included in these kits has been tested to ensure a low background for infrared Western blot success. This kit contains Intercept (TBS) Blocking Buffer (500 mL) and Millipore Immobilon-FL PVDF Membrane (1 roll; 0.45µm, 26.5 cm x 3.75 m).
Catalog No. NC1717883