Learn More
Electron Microscopy Sciences Veco Single Hole Grid A600, Copper, 600 µm hole
SDP
Supplier:  Electron Microscopy Sciences 0600CU
Encompass_Preferred
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter .
Material: Copper
Hole: 600 µm
Catalog No. 50-980-130