Learn More
Alkali Scientific Single ServeFetal Bovine Serum
SDP
Supplier:  Alkali Scientific FB72S
Encompass
CellProSingle ServeFBS is an aliquot-free solution for serum use minimizing contamination and saving time The Single ServeFBS 50 mL bottle is designed to eliminate a tedious lab task and significantly improve your cell culture results By choosing Single ServeFBS a typical customer using 20 L of FBS per year would gain almost 7 hours back and generate 7 lb less plastic waste This is a significant benefit both to lab productivity and the environment CellProFetal Bovine Serum (FBS) is the liquid part of the blood depleted of the cellular fraction and coagulation factors It is extracted after centrifugation of coagulated fetal bovine blood collected aseptically Sterile FBS is obtained by triple filtration of 0 1 m FBS contains proteins and growth factors allowing maintenance and proliferation of eukaryote cells in vitro It also contains attachment factors that create a coating effect on the plastic of the support thus increasing cell adhesion It is an essential product in cell cul
Catalog No. 50-302-7362