Learn More
Alkali Scientific Single ServeFetal Bovine Serum
SDP
Supplier:  Alkali Scientific FB73SCS
Encompass_Preferred
CellProSingle ServeFBS is an aliquot-free solution for serum use minimizing contamination and saving time The Single ServeFBS 50 mL bottle is designed to eliminate a tedious lab task and significantly improve your cell culture results By choosing Single ServeFBS a typical customer using 20 L of FBS per year would gain almost 7 hours back and generate 7 lb less plastic waste This is a significant benefit both to lab productivity and the environment CellProPremium Imported (USDA Approved) FBS is ideal for the most sensitive cell lines and can be used to culture all cell types including macrophages cancer and cancer related cells Hybridomas and more It works great for all our human primary cells and stem cells Our Alkali ScientificFBS is manufactured in a fully integrated environment from raw material collection to finished product It is produced to the highest standards in order to reduce risk and product variability and to ensure product safety and performance Triple 0 1 m steri
Catalog No. 50-302-6914