Learn More
Electron Microscopy Sciences Veco Grid L0.2 x 1.5, Nickel with 200 x 1500 µm Oval Slot
SDP
Supplier:  Electron Microscopy Sciences 0215NI
Encompass_Preferred
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter.
Slot: 200 x 1500 µm
Material: Nickel
Catalog No. 50-980-119