Learn More
BURKLE INC DispoPipette SteriPlast, HDPE sterile,39.37 in. (1000 mm), 6.76 oz. (200 ml)
Supplier:  BURKLE INC 53935532
Encompass_Preferred
Simple disposable sampler for taking samples of low viscosity media. If the DispoPipette is inserted into the liquid opened, an all-layer sample is obtained. If it is closed while it is inserted up to the sampling point and then opened, a target-point sample is obtained. Very easy to operate. With two opening sizes (2.5/5.5 mm (0.10/0.22 in.)) for different viscosities.
Catalog No. 50-308-1179