Learn More
BURKLE INC ZoneDispo LaboPlast, HDPE, 19.69 in. (500 mm), 3.38 oz. (100 ml)
Supplier:  BURKLE INC 53934420
Encompass_Preferred
The ZoneDispo disposable all-layer sampler is used in sampling to collect a representative all-layer sample from three different layers. Suitable for free-flowing powders and small-grain granulates. The collected sample can be secured and transported in the closed sampler.
Catalog No. 50-308-1422