Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Conductive Black Single Wafer Container, 2.5", Polypropylene Material, 75 x 11 mm
SDP

Supplier:  Electron Microscopy Sciences 6491925

Encompass_Preferred

The carriers are used for storage and or shipping wafers. They prevent all types of damage and contamination to the wafer during shipping and even storage. Each of the containers are packed in 1000 class clean room and bag. The set includes the Container , the cover and the spring.
Use: 2.5" (63mm)
Material: Polypropylene
Dimensions: 75 x 11 mm

Catalog No. 50-192-9907


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.