Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Single-Use Tying Angled Forceps, 6.5 mm tying platform Tips, 80 mm Handle
SDP

Supplier:  Electron Microscopy Sciences 7793792

Encompass_Preferred

These unique sterile forceps have a plastic base and tips made of are 316 stainless steel.
Tips: 6.5 mm tying platform
Handle: 80 mm
OAL: 100 mm

Catalog No. 50-193-1094


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.