Learn More
Foxx Life Sciences EZBio Dip Tube Sinker, PVDF, 1/8" Barb, Individually Double Bagged, Non-Sterile, 1/EA
Supplier:  Foxx Life Sciences 25S1228FLS
Encompass_Preferred
Foxx Life Sciences offers a premium line of high-quality PVDF Anchor dip tube sinkers. The Anchor is developed with you in mind to help eliminate the irritation of moving dip tubes. With this unique design, the Anchor helps to ensure optimal depth and flow rate, while removing the risk of suction against all containers. SpecificationsAnchor Material: PVDFBarb Size: 1/8"Part Diameter: 0.75"Gamma Irradiated: NoQuantity: 1 eachPackaging: Individually Double Bagged
Catalog No. 50-206-0598