Learn More
Apexbio Technology LLC Sinomenine 115-53-7 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC N17225.1
Encompass_Preferred
Sinomenine (CAS 115-53-7) is an alkaloid derived from the leaves of Sinomenium acutum It is known to modulate multiple inflammatory pathways including inhibition of NF- B activation and suppression of pro-inflammatory cytokine production Sinomenine also exhibits immunosuppressive effects by affecting T cell and macrophage function Owing to these properties it has been extensively used in research focused on autoimmune disorders inflammatory diseases and pain modulation The compound is soluble in ethanol acetone chloroform dilute alkali and hot benzene and should be stored in sealed cool and dry conditions to maintain stability
Catalog No. 50-000-90677