Learn More
Medchemexpress LLC HY-15122A 100mg Medchemexpress, Sinomenine hydrochloride CAS:6080-33-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY15122A100MG
Encompass_Preferred
Medchemexpress, HY-15122A 100mg Sinomenine hydrochloride CAS:6080-33-7 Sinomenine hydrochloride (Cucoline hydrochloride), an alkaloid extracted from Sinomenium acutum, is a blocker of the NF-κB activation. Sinomenine also is an activator of μ-opioid receptor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1681