Learn More
Medchemexpress LLC Sinomenine N-oxide | 1000026-77-6 | ≥99.0% | 345.39 g·mol-1 | C19H23NO5 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY15124110MG
Encompass_Preferred
Sinomenine N-oxide is a naturally derived alkaloid research reagent that exhibits anti-angiogenic, anti-inflammatory, and anti-rheumatic activities. It acts as a nitric oxide production inhibitor (reported IC50 23.04 μM) and is used in studies of NO synthase-related pathways in immunology and inflammation.
  • Inhibits nitric oxide production (IC50 23.04 μM).
  • Displays anti-angiogenic, anti-inflammatory, and anti-rheumatic activity.
  • Chemical formula C19H23NO5; molecular weight 345.39 g·mol-1.
  • Provided as a high-purity research compound (≥99.0%).
  • Suitable for cellular assays targeting NO synthase and inflammation pathways.
  • Available in small research quantities such as 10 MG.
Catalog No. 50-002-86922