Learn More
Medchemexpress LLC Sinomenine N-oxide | 1000026-77-6 | C19H23NO5 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1512415MG
Encompass_Preferred
Sinomenine N-oxide exhibits anti-angiogenic, anti-inflammatory, and anti-rheumatic effects. It functions as a nitric oxide (NO) production inhibitor, demonstrating an IC50 value of 23.04 μM.
  • Inhibits IL-6 and TNF-α levels in LPS-induced Raw264.7 cells.
Catalog No. 50-003-74803