Learn More
Perkin Elmer US LLC Sipper Cell and Pump Accessory
Supplier:  Perkin Elmer US LLC L2250135
Encompass_Preferred
The Sipper Cell and Pump Accessory measures large numbers of liquid samples quickly and accurately. Samples can be transferred from a wide variety of containers to the flow cell. Includes: Synthetic Fused Silica flow cell, pathlength: 1.3 mm, cell volume: 16 mL (L2151247) Peristaltic pump controlled by computer Standard fitting 0.4 mm FEP Inlet Tube (L2151202) 0.7 mm FEP Outlet Tube (L2151203) Please note: This accessory requires installation by a PerkinElmer service engineer, not included.
Catalog No. 50-211-2755