Learn More
Selleck Chemical LLC Siramesine HCl
SDP
Supplier:  Selleck Chemical LLC S67562MG
Encompass_Preferred
Siramesine (Lu 28-179) is a sigma receptor agonist selective for the 2 subtype with IC50 values of 0 12 nM and 17 nM for 2 and 1 receptors respectively
Catalog No. 50-000-02899