Learn More
Medchemexpress LLC Siramesine (hydrochloride) | 224177-60-0 | 99.7% | C30H32ClFN2O | 1 ML
SDP
Supplier:  Medchemexpress LLC HY14221A10MM/1ML
Encompass_Preferred
Siramesine hydrochloride is a potent sigma-2 receptor agonist with subnanomolar affinity (IC50=0.12 nM), showing significant selectivity over sigma-1 receptors. This compound triggers cell death by destabilizing mitochondria and demonstrates anti-cancer activity.
  • Potent sigma-2 receptor agonist
  • Subnanomolar affinity for sigma-2 receptors
  • High selectivity over sigma-1 receptors
  • Triggers cell death through mitochondrial destabilization
  • Exhibits anti-cancer activity
  • Induces cell death in various cell lines
  • Activates caspases in HaCaT and U-87MG cells
Catalog No. 50-003-53609