Learn More
Medchemexpress LLC HY-18658 1mg , Siremadlin CAS:1448867-41-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY186581MG
Encompass_Preferred
HY-18658 1mg Siremadlin CAS: 1448867-41-1 Siremadlin (NVP-HDM201) is a potent, orally bioavailable and highly specific p53-MDM2 interaction inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9558