Learn More
TARGETMOL CHEMICALS INC SIREMADLIN 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T55555MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Siremadlin (NVP-HDM 201) is a potent orally bioavailable and highly specific p53-MDM2 interaction inhibitor. purity: 98%
Catalog No. 50-257-6674