Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sirolimus (rapamycin) | 53123-88-9 | MFCD00867594 | 99.0% | 914.17 | C51H79NO13 | 10 MG
SDP

Supplier:  Medchemexpress LLC HY10219R10MG

Encompass_Preferred

Rapamycin (sirolimus) analytical standard supplied as a 10 mg reference material for research and analytical applications. It is intended for use in assay calibration, method validation, and analytical reference testing; for research use only.

  • Analytical standard for rapamycin (sirolimus).
  • Useful for assay calibration and method validation.
  • Molecular formula C51H79NO13.
  • Molecular weight 914.17.
  • Supplied as a 10 MG reference quantity.
  • Intended for research and analytical applications; not for clinical use.

Catalog No. 50-002-47063


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.