Learn More
Medchemexpress LLC SIRP alpha/CD172a H 100ug | 100ug
SDP
Supplier:  Medchemexpress LLC HYP78211100UG
Encompass_Preferred
The SIRP V4/CD172a protein is an immunoglobulin-like cell surface receptor for CD47 that acts as a docking protein to facilitate the translocation of PTPN6 PTPN11 and other partners to the plasma membrane It supports the adhesion of cerebellar neurons promotes neurite growth and promotes glial cell attachment SIRP alpha/CD172a Protein Human (Biotinylated HEK293 Fc-Avi) is the recombinant human-derived SIRP alpha/CD172a protein expressed by HEK293 with C-Avi C-hFc labeled tag
Catalog No. 50-001-93778