Learn More
Medchemexpress LLC Sirt-in-2 | 1431411-66-3 | 98.1% | 383.49 | C15H21N5O3S2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY1661610MG
Encompass_Preferred
Sirt-in-2 is a potent, low-nanomolar inhibitor of sirtuin deacetylases SIRT1, SIRT2, and SIRT3 intended for laboratory research. The compound is provided as a high-purity solid (typical purity 98.1%) in a milligram pack for preparing stock solutions; it is for research use only and not for human use.
  • Inhibits SIRT1, SIRT2, and SIRT3 with IC50s of 4 nM, 1 nM, and 7 nM, respectively.
  • High purity suitable for biochemical and cell-based assays (≈98.1%).
  • Molecular formula C15H21N5O3S2 and molecular weight 383.49.
  • Supplied as a milligram-scale solid (e.g., 10 mg) for preparing stock solutions.
  • Intended for research use only; not for human or clinical use.
Catalog No. 50-002-88566