Learn More
Abcam SIRT inhibitor 1/2 VII, 10MG
Supplier:  Abcam AB14455010MG
Encompass_Preferred
MW 282.3 Da, Purity >98%. Sirtuin and NAD+-dependent class III HDAC inhibitor (IC₅₀ values are 13.2 and 9.1 μM for SIRT1 and SIRT2, respectively). Thiobarbiturate compound.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5637