Learn More
Medchemexpress LLC SIRT1-IN-3 | 2470969-91-4 | 99.1% | 295.18 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY14669010MM/1ML
Encompass_Preferred
SIRT1-IN-3 (compound 3j) is a potent and selective SIRT1 inhibitor with an IC50 of 4.2 μM. It inhibits the proliferation of several human cancer cell lines, including K562, HCT-116, H460, HepG2, A549, and MCF-7, while demonstrating low cytotoxicity on 293T and HUVEC.
  • Potent and selective SIRT1 inhibitor
  • Inhibits proliferation of human cancer cell lines
  • Low cytotoxicity on 293T and HUVEC
  • Available as a 10 mM, 1 mL solution in DMSO
Catalog No. 50-003-62905