Learn More
Medchemexpress LLC SIRT5 inhibitor 1 | 2166487-21-2 | 99.7% | 674.81 | 5 MG

Supplier: Medchemexpress LLC 5MGHY112634
SIRT5 inhibitor 1 is a potent human sirtuin 5 deacylase inhibitor, with an IC50 of 0.11 μM. This compound is described as a very potent human sirtuin 5 deacylase inhibitor, with an IC50 of 0.11 μM, >100-fold from compound 1.
- Potent human sirtuin 5 deacylase inhibitor
- IC50 of 0.11 μM
- Target: Sirtuin
- Pathways: Cell Cycle/DNA Damage; Epigenetics
- For research use only
- Appearance: White to off-white solid
- Molecular formula: C31H39FN6O6S2
- Storage powder: -20°C for 3 years, 4°C for 2 years
- Storage in solvent: -80°C for 2 years, -20°C for 1 year
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.