Learn More
TARGETMOL CHEMICALS INC SIRT5 INHIBITOR 1 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T129215MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SIRT5 inhibitor 1 is a potent and selective inhibitor of human Sirtuin 5 deacylase (IC50 0.11 uM). sIRT5 inhibitor 1 is associated with ageing-related diseases. purity: 98%
Catalog No. 50-258-1398