Learn More
Apexbio Technology LLC Sirtinol, 5mg. Cas: 410536-97-9 MFCD: MFCD00810186. SIRT inhibitor
SDP
Supplier:  Apexbio Technology LLC A41835
Encompass_Preferred
Sirtinol is an inhibitor of SIRT with IC50 value of 48.6 M (24 h) and 43.5 M (48 h) in MCF-7 cells [1].Sirtinol significantly reduces the growth of MCF-7 cells in a concentration- and time-dependent manner. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4900